Tisk…
tert-Butyllithium ∼1.9M in pentane
Dodavatel: Thermo Scientific
Nebezpečí
Synonyms: t-Butyllithium
Packaged under Nitrogen in resealable AcroSeal® bottles
H36881.AE H36881.9Z
H36881.AEEA 3050 CZK
H36881.AE H36881.9Z
tert-Butyllithium ∼1.9M in pentane
tert-Butyllithium
Vzorec: C₄H₉Li
MW: 64,06 g/mol
Bod vzplanutí: –49°C (–56°F)
Storage Temperature: Ambient
MDL Number: MFCD00008795
CAS číslo: 594-19-4
EINECS: 209-831-5
UN: 3394
ADR: 4.2,I