n-Butyllithium 1.6M in hexanes
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Packaged under Nitrogen in resealable AcroSeal® bottles
H36033.9Z
H36033.AE
H36033.AP
H36033.9ZEA
6010
CZK
H36033.9Z
H36033.AE
H36033.AP
n-Butyllithium 1.6M in hexanes
n-Butyllithium
Vzorec:
C₄H₉Li MW: 64,06 g/mol Bod vzplanutí: –21°C (–6°F) Storage Temperature: Ambient |
MDL Number:
MFCD00009414 CAS číslo: 109-72-8 EINECS: 203-698-7 UN: 3394 ADR: 4.2,I |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...