Tisk…

Diisobutylaluminium hydride 1.1M in cyclohexane

Dodavatel: Thermo Scientific
Nebezpečí


Packaged under Nitrogen in resealable AcroSeal® bottles

H37108.9Z H37108.AE
H37108.9ZEA 11840 CZK
H37108.9Z H37108.AE
Diisobutylaluminium hydride 1.1M in cyclohexane
Diisobutylaluminium hydride
Vzorec: C₈H₁₉Al
MW: 142,22 g/mol
Bod vzplanutí: –18°C (0°F)
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS číslo: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR