Tisk…

Adamantane-1-carboxylic acid ≥99%

Dodavatel: Thermo Scientific
Varování

A12959.14 A12959.22 A12959.36
A12959.14EA 1800 CZK
A12959.14 A12959.22 A12959.36
Adamantane-1-carboxylic acid ≥99%
Adamantane-1-carboxylic acid

Purity: 99% CAS No.: 828-51-3 Molecular Formula: C11H16O2 Formula Weight: 180.25 Melting Point: 173-176° Beilstein Reference: 1910637

Vzorec: C₁₁H₁₆O₂
MW: 180,25 g/mol
Bod tání: 173…176 °C
Hustota: 1,221 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074720
CAS číslo: 828-51-3
EINECS: 212-584-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR