Melamin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine
M-Clarity™ quality level = MQ200
M2659-100G
M2659-1KG
M2659-3KG
M2659-5G
SIALM2659-100GEA
1240
CZK
SIALM2659-100G
SIALM2659-1KG
SIALM2659-3KG
SIALM2659-5G
Melamin, Sigma-Aldrich®
Melamin
Vzorec:
C₃H₆N₆ MW: 126,12 g/mol Hustota: 1,57 g/cm³ (20 °C) Bod vzplanutí: >280 °C |
MDL Number:
MFCD00006055 CAS číslo: 108-78-1 EINECS: 203-615-4 Merck Index: 13,05834 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...