Melamin for synthesis, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine
M-Clarity™ quality level = MQ200
8.08614.0100
8.08614.1000
8.08614.0100EA
835
CZK
8.08614.0100
8.08614.1000
Melamin for synthesis, Sigma-Aldrich®
Melamin
Vzorec:
C₃H₆N₆ Hustota: 1,57 g/cm³ (20 °C) Bod vzplanutí: >280 °C |
MDL Number:
MFCD00006055 CAS číslo: 108-78-1 EINECS: 203-615-4 Merck Index: 13,05834 |
Výsledky specifikačního testu
Solubility | 3.2 g/l (20 °C) |
Melting Point | 354 °C (decomposition) |
Molar Mass | 126.12 g/mol |
Bulk Density | 800 kg/m3 |
Boiling Point |
|
Vapor Pressure | 66.65 hPa (315 °C) |
Density | 1.57 g/cm3 (20 °C) |
pH | 7 - 8 (32 g/l, H2O, 20 °C) |
Ignition Point |
|
Synonyms | Melamine |
Hill Formula | C3H6N6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...