2,4,6-Trifluoro-1,3,5-triazine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A15666.09
A15666.22
A15666.09EA
0
CZK
A15666.09
A15666.22
2,4,6-Trifluoro-1,3,5-triazine ≥98%
2,4,6-Trifluoro-1,3,5-triazine
Purity: 98%
CAS No.: 675-14-9
Molecular Formula: C3F3N3
Formula Weight: 135.05
Refractive Index: 1.3840
Odour: Acrid
Boiling Point: 73-74°
Beilstein Reference: 124237
UN No.: 3389
MDL No.: MFCD00014597
Vzorec:
C₃F₃N₃ Bod varu: 73…74 °C Bod tání: –38 °C Hustota: 1,574 Storage Temperature: Ambient |
MDL Number:
MFCD00014597 CAS číslo: 675-14-9 EINECS: 211-620-8 UN: 3389 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...