Cyanuric acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid
M-Clarity™ quality level = MQ200
185809-25G
185809-100G
SIAL185809-25GEA
1270
CZK
SIAL185809-25G
SIAL185809-100G
Cyanuric acid, Sigma-Aldrich®
Cyanuric acid
Vzorec:
C₃H₃N₃O₃ MW: 129,08 g/mol Hustota: 2,5 g/cm³ (20 °C) |
MDL Number:
MFCD00082990 CAS číslo: 108-80-5 EINECS: 203-618-0 Merck Index: 13,02728 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...