Cyanuric acid for synthesis, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid
M-Clarity™ quality level = MQ200
8.20358.0005
8.20358.0250
8.20358.1000
8.20358.0005EA
790
CZK
8.20358.0005
8.20358.0250
8.20358.1000
Cyanuric acid for synthesis, Sigma-Aldrich®
Cyanuric acid
Vzorec:
C₃H₃N₃O₃ Hustota: 2,5 g/cm³ (20 °C) |
MDL Number:
MFCD00082990 CAS číslo: 108-80-5 EINECS: 203-618-0 Merck Index: 13,02728 |
Výsledky specifikačního testu
Solubility | 2 g/l (20 °C) |
Melting Point | 320 - 330 °C (decomposition) |
Molar Mass | 129.07 g/mol |
Bulk Density | 550 kg/m3 |
Vapor Pressure |
|
Density | 2.5 g/cm3 (20 °C) |
pH | 3.8 - 4.0 (H2O, 20 °C) (saturated solution) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...