Tisk…

Cyanuric acid ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: 1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid

A15447.0B A15447.22 A15447.30
A15447.0BEA 3810 CZK
A15447.0B A15447.22 A15447.30
Cyanuric acid ≥99%
Cyanuric acid

Purity: 99% CAS No.: 108-80-5 Molecular Formula: C3H3N3O3 Formula Weight: 129.08 Melting Point: 360° Beilstein Reference: 126982 Merck Reference: 14,2698 MDL No.: MFCD00082990

Vzorec: C₃H₃N₃O₃
Bod tání: >360 °C
Hustota: 2,5
Storage Temperature: Ambient
MDL Number: MFCD00082990
CAS číslo: 108-80-5
EINECS: 203-618-0
Merck Index: 14,2698

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR