Tetrahydrothiophene, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
THT
M-Clarity™ quality level = MQ100
T15601-100G
T15601-500G
T15601-5G
SIALT15601-100GEA
1360
CZK
SIALT15601-100G
SIALT15601-500G
SIALT15601-5G
Tetrahydrothiophene, Sigma-Aldrich®
Tetrahydrothiophene
Vzorec:
C₄H₈S MW: 88,17 g/mol Bod varu: 119 °C (1013 hPa) Bod tání: –96 °C Hustota: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00005476 CAS číslo: 110-01-0 EINECS: 203-728-9 UN: 2412 ADR: 3,II Merck Index: 12,09357 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...