2-Picoline-N-oxide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Methylpyridine-N-oxide, 2-Picoline N-oxide, 2-Methylpyridine-1-oxide, 2-Methylpyridine 1-oxide
L04488.14
L04488.22
L04488.36
L04488.14EA
951
CZK
L04488.14
L04488.22
L04488.36
2-Picoline-N-oxide ≥98%
2-Picoline-N-oxide
Purity: 98%
CAS No.: 931-19-1
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 143°(289°F)
Melting Point: 46-50°
Boiling Point: 122°/8mm
Beilstein Reference: 106916
MDL No.: MFCD00006199
Vzorec:
C₆H₇NO MW: 109,13 g/mol Bod varu: 122 °C (8 mmHg) Bod tání: 46…50 °C Bod vzplanutí: 143 °C (289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006199 CAS číslo: 931-19-1 EINECS: 213-230-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...