3-Hydroxy-2-nitropyridine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Nitropyridin-3-ol, 2-Nitro-3-pyridinol
M-Clarity™ quality level = MQ100
107255-100G
107255-25G
SIAL107255-100GEA
11400
CZK
SIAL107255-100G
SIAL107255-25G
3-Hydroxy-2-nitropyridine, Sigma-Aldrich®
3-Hydroxy-2-nitropyridine
Vzorec:
C₅H₄N₂O₃ MW: 140,1 g/mol Bod tání: 70 °C |
MDL Number:
MFCD00006258 CAS číslo: 15128-82-2 EINECS: 239-191-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...