Tisk…

2-Chloro-4-nitropyridine-N-oxide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

534269-25G 534269-5G
SIAL534269-25GEA 6870 CZK
SIAL534269-25G SIAL534269-5G
2-Chloro-4-nitropyridine-N-oxide, Sigma-Aldrich®
2-Chloro-4-nitropyridine-N-oxide
Vzorec: C₅H₃ClN₂O₃
MW: 174,54 g/mol
Bod tání: 158…159 °C
MDL Number: MFCD00955617
CAS číslo: 14432-16-7
EINECS: 238-404-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR