2-Bromo-3-fluoropyridine ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H31981.03
H31981.06
H31981.03EA
1370
CZK
H31981.03
H31981.06
2-Bromo-3-fluoropyridine ≥97%
2-Bromo-3-fluoropyridine
Purity: 97%
CAS No.: 40273-45-8
Molecular Formula: C5H3BrFN
Formula Weight: 175.99
Refractive Index: 1.5450
Vzorec:
C₅H₃BrFN Storage Temperature: Ambient |
MDL Number:
MFCD04114128 CAS číslo: 40273-45-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...