2-Acetyl-3-methylpyrazine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W396400-100G-K
W396400-1KG-K
W396400-25G-K
SIALW396400-100G-KEA
9590
CZK
SIALW396400-100G-K
SIALW396400-1KG-K
SIALW396400-25G-K
2-Acetyl-3-methylpyrazine, Sigma-Aldrich®
2-Acetyl-3-methylpyrazine
Vzorec:
C₇H₈N₂O Bod varu: 90 °C (1013 hPa) Hustota: 1,11 g/cm³ (20 °C) |
MDL Number:
MFCD00014612 CAS číslo: 23787-80-6 EINECS: 245-889-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...