2-Methoxy-3-methylpyrazine ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L08067.06
L08067.14
L08067.06EA
1730
CZK
L08067.06
L08067.14
2-Methoxy-3-methylpyrazine ≥99%
2-Methoxy-3-methylpyrazine
Purity: 99%
CAS No.: 2847-30-5
Molecular Formula: C6H8N2O
Formula Weight: 124.14
Flash Point: 55°(131°F)
Refractive Index: 1.5065
Boiling Point: 159°
Beilstein Reference: 878809
UN No.: 1993
MDL No.: MFCD00006127
Vzorec:
C₆H₈N₂O MW: 124,14 g/mol Bod varu: 159 °C Hustota: 1,070 Bod vzplanutí: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006127 CAS číslo: 2847-30-5 EINECS: 220-651-6 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...