Tisk…

2-Ethyl-3-methoxypyrazine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W514403-100G-K W514403-25G-K
SIALW514403-100G-KEA 20590 CZK
SIALW514403-100G-K SIALW514403-25G-K
2-Ethyl-3-methoxypyrazine, Sigma-Aldrich®
2-Ethyl-3-methoxypyrazine
Vzorec: C₇H₁₀N₂O
MW: 138,17 g/mol
Bod varu: 81…85 °C (20 mmHg)
Hustota: 1,04 g/cm³ (20 °C)
MDL Number: MFCD00006129
CAS číslo: 25680-58-4
EINECS: 247-184-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR