Pyrazine-2-carboxylic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Pyrazinecarboxylic acid, 2-Pyrazinecarboxylic Acid
M-Clarity™ quality level = MQ200
P56100-100G
P56100-25G
SIALP56100-100GEA
3550
CZK
SIALP56100-100G
SIALP56100-25G
Pyrazine-2-carboxylic acid, Sigma-Aldrich®
Pyrazine-2-carboxylic acid
Vzorec:
C₅H₄N₂O₂ MW: 124,1 g/mol Bod varu: 313 °C (1013 hPa) |
MDL Number:
MFCD00006130 CAS číslo: 98-97-5 EINECS: 202-718-1 Merck Index: 12,08143 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...