Pyrazine-2-carboxylic acid 99%
Dodavatel: Thermo Scientific
Synonyms:
Pyrazinecarboxylic acid, 2-Pyrazinecarboxylic Acid
131710250
131711000
ACRO131710250EA
1110
CZK
ACRO131710250
ACRO131711000
Pyrazine-2-carboxylic acid 99%
Pyrazine-2-carboxylic acid
Vzorec:
C₅H₄N₂O₂ MW: 124,1 g/mol Bod varu: 313 °C (1013 hPa) Bod tání: 222…225 °C |
MDL Number:
MFCD00006130 CAS číslo: 98-97-5 EINECS: 202-718-1 Merck Index: 12,08143 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 222 to 225 °C |
Titration with NaOH | ≥98.5 % |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...