Methylene blue trihydrate, pure, certified residual water
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3,7-Bis(dimethylamino)phenothiazin-5-ium chloride trihydrate, C.I.52015, Methylthioninium chloride trihydrate, Basic blue 9, Swiss Blue, Methylene blue
414240250
414241000
414245000
ACRO414240250EA
1930
CZK
ACRO414240250
ACRO414241000
ACRO414245000
Methylene blue trihydrate, pure, certified residual water
Methylene blue trihydrate
Vzorec:
C₁₆H₁₈ClN₃S·3H₂O MW: 373,9 g/mol Bod tání: 100…110 °C Hustota: 1,757 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012111 CAS číslo: 7220-79-3 EINECS: 200-515-2 Merck Index: 12,06137 |
Výsledky specifikačního testu
Appearance | Dark green Crystalline powder |
Infrared spectrum | Conforms |
Dye content | ≥82 % (as is basis) |
UV Lambda max | 663 to 667 nm (in water) |
Absorption Ratio P-15/P+15 | 1.21 to 1.70 |
BSC Certification | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...