Maltol, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone
M-Clarity™ quality level = MQ300
W265608-1KG-K
W265608-250G-K
W265608-5KG-K
SIALW265608-1KG-KEA
3030
CZK
SIALW265608-1KG-K
SIALW265608-250G-K
SIALW265608-5KG-K
Maltol, Sigma-Aldrich®
Maltol
Vzorec:
C₆H₆O₃ MW: 126,11 g/mol Bod varu: 285 °C (1013 hPa) Hustota: 1,453 g/cm³ (20 °C) |
MDL Number:
MFCD00006578 CAS číslo: 118-71-8 EINECS: 204-271-8 Merck Index: 13,05735 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...