Tisk…

Maltol ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone

A10735.14 A10735.22
A10735.14EA 1350 CZK
A10735.14 A10735.22
Maltol ≥99%
Maltol

Purity: 99% CAS No.: 118-71-8 Molecular Formula: C6H6O3 Formula Weight: 126.11 Flash Point: 95°(203°F) Melting Point: 161-165° Boiling Point: 170°/100mm Beilstein Reference: 112169 Merck Reference: 14,5713 MDL No.: MFCD00006578

Vzorec: C₆H₆O₃
MW: 126,11 g/mol
Bod varu: 170 °C (100 mmHg)
Bod tání: 161…165 °C
Hustota: 1,453 g/cm³ (20 °C)
Bod vzplanutí: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006578
CAS číslo: 118-71-8
EINECS: 204-271-8
Merck Index: 14,5713

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR