Tisk…

Cobalt (III) acetylacetonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

C83902-100G C83902-25G
SIALC83902-100GEA 6410 CZK
SIALC83902-100G SIALC83902-25G
Cobalt (III) acetylacetonate, Sigma-Aldrich®
Cobalt (III) acetylacetonate
Vzorec: C₁₅H₂₁CoO₆
MDL Number: MFCD00013488
CAS číslo: 21679-46-9
EINECS: 244-527-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR