Cobalt (III) acetylacetonate
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Cobalt (III) tris(2,4-pentanedionato-O,O'), Cobalt (III) 2,4-pentanedionate, Cobalt tris(acetylacetonate), Tris(2,4-pentanedionato-O,O')cobalt (III)
12536.06
12536.14
12536.22
12536.06EA
1220
CZK
12536.06
12536.14
12536.22
Cobalt (III) acetylacetonate
Cobalt (III) acetylacetonate
CAS No.: 21679-46-9
Molecular Formula: C15H21CoO6
Formula Weight: 356.26
Physical Form: Powder
Sensitivity: Moisture Sensitive and Hygroscopic
Odour: Light
MDL No.: MFCD00013488
Vzorec:
C₁₅H₂₁CoO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00013488 CAS číslo: 21679-46-9 EINECS: 244-527-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...