Tisk…

Platinum(II) acetylacetonate (≥48,0% Pt)

Dodavatel: Thermo Scientific
Varování

Synonyms: Platinum (II) bis(2,4-pentanedionato-O,O'), Platinum (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')platinum (II)

10526.03 10526.06
10526.03EA 6320 CZK
10526.03 10526.06
Platinum(II) acetylacetonate (≥48,0% Pt)
Platinum(II) acetylacetonate

CAS No.: 15170-57-7 Molecular Formula: C10H14O4Pt Formula Weight: 393.31 Physical Form: Needles Appearance: Pale yellow Melting Point: 250-252° MDL No.: MFCD00000028

Vzorec: C₁₀H₁₄O₄Pt
MW: 393,3 g/mol
Bod tání: 250…252 °C
Hustota: ∼1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000028
CAS číslo: 15170-57-7
EINECS: 239-223-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR