Tisk…

Palladium(II) acetylacetonate (35% Pd)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Palladium (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')palladium (II), Palladium (II) bis(2,4-pentanedionato-O,O'), Palladium(II) 2,4-pentanedionate

197580010 197580050
ACRO197580010EA 4120 CZK
ACRO197580010 ACRO197580050
Palladium(II) acetylacetonate (35% Pd)
Palladium(II) acetylacetonate
Vzorec: C₁₀H₁₄O₄Pd
MW: 304.64
Bod tání: 190 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00000025
CAS číslo: 14024-61-4
EINECS: 237-859-8

Objednat

Výsledky specifikačního testu

Appearance Yellow to green Powder
Infrared spectrum Conforms
Assay 33.9 to 35.9 % (Pd)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR