Tisk…

Nickel(II) acetylacetonate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Bis(2,4-pentanedionato-O,O')nickel (II), Nickel (II) bis(2,4-pentanedionato-O,O'), Nickel acetylacetonate, Nickel (II) 2,4-pentanedionate

M-Clarity™ quality level = MQ100

283657-25G 283657-50G 283657-5G
SIAL283657-25GEA 6140 CZK
SIAL283657-25G SIAL283657-50G SIAL283657-5G
Nickel(II) acetylacetonate, Sigma-Aldrich®
Nickel(II) acetylacetonate
Vzorec: C₁₀H₁₄NiO₄
MW: 256,91 g/mol
Bod varu: 220 °C (1013 hPa)
Bod tání: 208 °C
Hustota: 1,455 g/cm³ (20 °C)
MDL Number: MFCD00000024
CAS číslo: 3264-82-2
EINECS: 221-875-7
Merck Index: 12,06584

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR