Tisk…

Nickel(II) acetylacetonate, anhydrous ≥95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Bis(2,4-pentanedionato-O,O')nickel (II), Nickel (II) bis(2,4-pentanedionato-O,O'), Nickel acetylacetonate, Nickel (II) 2,4-pentanedionate

18811.06 18811.14 18811.22 18811.36
18811.06EA 968 CZK
18811.06 18811.14 18811.22 18811.36
Nickel(II) acetylacetonate, anhydrous ≥95%
Nickel(II) acetylacetonate

Purity: 95% CAS No.: 3264-82-2 Molecular Formula: C10H14NiO4 Formula Weight: 256.91 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Green Odour: Odourless Melting Point: ca 230° dec. Solubility: Soluble in water, alcohol, toluene Beilstein Reference: 4157970 Merck Reference: 14,6500 MDL No.: MFCD00000024


Catalyst and U.V. stabilizer

Vzorec: C₁₀H₁₄NiO₄
Bod varu: 220 °C (1013 hPa)
Hustota: 1,455 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000024
CAS číslo: 3264-82-2
EINECS: 221-875-7
Merck Index: 14,6500

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR