Tisk…

Nickel(II) acetylacetonate 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Bis(2,4-pentanedionato-O,O')nickel (II), Nickel (II) bis(2,4-pentanedionato-O,O'), Nickel acetylacetonate, Nickel (II) 2,4-pentanedionate

128260250 128260050 128261000
ACRO128260250EA 2370 CZK
ACRO128260250 ACRO128260050 ACRO128261000
Nickel(II) acetylacetonate 96%
Nickel(II) acetylacetonate
Vzorec: C₁₀H₁₄NiO₄
MW: 256.91
Bod varu: 220 °C (11 mmHg)
Bod tání: 230 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00000024
CAS číslo: 3264-82-2
EINECS: 221-875-7
Merck Index: 12,06584

Objednat

Výsledky specifikačního testu

Appearance Green Powder
Infrared spectrum Conforms
Titration Complexometric ≥95 %
Chloride (Cl) ≤5 ppm
Phosphate (PO4) ≤5 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤10 ppm
Sodium (Na) ≤20 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR