Cobalt(II) acetylacetonate
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Cobalt(II) bis(acetylacetonate), Cobalt (II) bis(2,4-pentanedionato-O,O'), Cobalt (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')cobalt (II)
12537.18
12537.30
12537.18EA
1980
CZK
12537.18
12537.30
Cobalt(II) acetylacetonate
Cobalt(II) acetylacetonate
CAS No.: 14024-48-7
Molecular Formula: C10H14CoO4
Formula Weight: 257.14
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Blue/purple
Odour: Odourless
Melting Point: 165-170°
MDL No.: MFCD00000014
Vzorec:
C₁₀H₁₄CoO₄ MW: 257.15 Bod tání: 165…170 °C Hustota: 0.83 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000014 CAS číslo: 14024-48-7 EINECS: 237-855-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...