Tisk…
Palladium(II) oxide monohydrate 99.9% (≥73% Pd) (metals basis)
Dodavatel: Thermo Fisher Scientific
Synonyms: Palladium(II) oxide hydrate, Palladium monoxide monohydrate
11041.04 11041.09
11041.04EA 19740 CZK
11041.04 11041.09
Palladium(II) oxide monohydrate 99.9% (≥73% Pd) (metals basis)
Palladium(II) oxide monohydrate
Purity: 99.9% CAS No.: 64109-12-2 Molecular Formula: PdO.H2O Formula Weight: 140.41 (122.40anhy) Physical Form: Powder Melting Point: dec. MDL No.: MFCD00011172
Vzorec: OPd·1H₂O
MW: 140.43
Bod tání: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00011172
CAS číslo: 64109-12-2
EINECS: 215-218-3
Merck Index: 14,6993