Rhodium(III) iodide (≥20,8% Rh) ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
11029.01
11029.03
11029.06
11029.01EA
4770
CZK
11029.01
11029.03
11029.06
Rhodium(III) iodide (≥20,8% Rh) ≥99.9% (metals basis)
Rhodium(III) iodide
Purity: 99.9%
CAS No.: 15492-38-3
Molecular Formula: RhI3
Formula Weight: 483.62
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Black
Odour: Odourless
MDL No.: MFCD00016311
Vzorec:
I₃Rh Bod varu: 100 °C (1013 hPa) Hustota: 6,4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016311 CAS číslo: 15492-38-3 EINECS: 239-521-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...