Tisk…

Iron(II) iodide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ferrous iodide, Iron diiodide

M-Clarity™ quality level = MQ100

400858-10G 400858-1G
SIAL400858-10GEA 13260 CZK
SIAL400858-10G SIAL400858-1G
Iron(II) iodide, Sigma-Aldrich®
Iron(II) iodide
Vzorec: FeI₂
MW: 309,66 g/mol
MDL Number: MFCD00016094
CAS číslo: 7783-86-0
EINECS: 232-031-2
UN: 3260
ADR: 8,III
Merck Index: 12,04097

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR