Tisk…

Iron(III) fluoride, anhydrous ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

11487.09 11487.18
11487.09EA 1490 CZK
11487.09 11487.18
Iron(III) fluoride, anhydrous ≥97%
Iron(III) fluoride

Purity: 97% min CAS No.: 7783-50-8 Molecular Formula: FeF3 Formula Weight: 112.84 Physical Form: Powder Density: 3.87 Appearance: Pale green Melting Point: 1000° subl. Solubility: Slightly soluble in water. Freely soluble in dilute HF. Insoluble in benzene, ether, alcohol Merck Reference: 14,4022 UN No.: 3260 MDL No.: MFCD00011012

Vzorec: F₃Fe
Bod tání: >1000 °C (subl.)
Hustota: 3,87
Storage Temperature: Ambient
MDL Number: MFCD00011012
CAS číslo: 7783-50-8
EINECS: 232-002-4
UN: 3260
ADR: 8,II
Merck Index: 14,4022

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR