Tisk…

Manganese(III) acetylacetonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

M2284-100G M2284-25G
SIALM2284-100GEA 5440 CZK
SIALM2284-100G SIALM2284-25G
Manganese(III) acetylacetonate, Sigma-Aldrich®
Manganese(III) acetylacetonate
Vzorec: C₁₅H₂₁MnO₆
Bod tání: 160 °C
MDL Number: MFCD00000023
CAS číslo: 14284-89-0
EINECS: 238-188-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR