Tisk…

Manganese(III) acetylacetonate

Dodavatel: Thermo Fisher Scientific

Synonyms: Manganese (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato-O,O')manganese (III), Manganese (III) 2,4-pentanedionate

12530.09 12530.18 12530.30
12530.09EA 1330 CZK
12530.09 12530.18 12530.30
Manganese(III) acetylacetonate
Manganese(III) acetylacetonate

CAS No.: 14284-89-0 Molecular Formula: C15H21MnO6 Formula Weight: 352.24 Physical Form: Powder Sensitivity: Hygroscopic Melting Point: ≈160° dec. MDL No.: MFCD00000023

Vzorec: C₁₅H₂₁MnO₆
MW: 352.27
Bod tání: ∼160 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00000023
CAS číslo: 14284-89-0
EINECS: 238-188-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR