Tisk…

Manganese(III) acetylacetonate 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Manganese (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato-O,O')manganese (III), Manganese (III) 2,4-pentanedionate

343980050 343980250 343981000
ACRO343980050EA 640 CZK
ACRO343980050 ACRO343980250 ACRO343981000
Manganese(III) acetylacetonate 97%
Manganese(III) acetylacetonate
Vzorec: C₁₅H₂₁MnO₆
Storage Temperature: Ambient
MDL Number: MFCD00000023
CAS číslo: 14284-89-0
EINECS: 238-188-3

Objednat

Výsledky specifikačního testu

Appearance Black Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR