Chromium(III) acetylacetonate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
202231-100G
202231-5G
SIAL202231-100GEA
1970
CZK
SIAL202231-100G
SIAL202231-5G
Chromium(III) acetylacetonate, Sigma-Aldrich®
Chromium(III) acetylacetonate
Vzorec:
C₁₅H₂₁CrO₆ Bod varu: 340 °C (1013 hPa) Hustota: 1,266 g/cm³ (20 °C) |
MDL Number:
MFCD00000015 CAS číslo: 21679-31-2 EINECS: 244-526-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...