Chromium(III) acetylacetonate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
12538.22
12538.36
12538.22EA
1690
CZK
12538.22
12538.36
Chromium(III) acetylacetonate ≥97%
Chromium(III) acetylacetonate
Purity: 97%
CAS No.: 21679-31-2
Molecular Formula: C15H21CrO6
Formula Weight: 349.33
Physical Form: Crystalline
Density: 1.35
Melting Point: 216°
Boiling Point: 340°
Solubility: Insoluble in water. Soluble in toluene, acetic acid
MDL No.: MFCD00000015
Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVD
Vzorec:
C₁₅H₂₁CrO₆ Bod varu: 340 °C Bod tání: 216 °C Hustota: 1,35 Storage Temperature: Ambient |
MDL Number:
MFCD00000015 CAS číslo: 21679-31-2 EINECS: 244-526-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...