Tisk…

Molybdenum pentachloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

208353-100G 208353-25G
SIAL208353-100GEA 7220 CZK
SIAL208353-100G SIAL208353-25G
Molybdenum pentachloride, Sigma-Aldrich®
Molybdenum pentachloride
Vzorec: Cl₅Mo
MW: 273.2
Bod varu: 268
Bod tání: 194
Hustota: 2.928 g/cm³ (20 °C)
MDL Number: MFCD00003467
CAS číslo: 10241-05-1
EINECS: 233-575-3
UN: 2508
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR