Molybdenum pentachloride ≥99.6% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
11832.06
11832.18
11832.30
11832.A1
11832.06EA
1150
CZK
11832.06
11832.18
11832.30
11832.A1
Molybdenum pentachloride ≥99.6% (metals basis)
Molybdenum pentachloride
Purity: 99.6%
CAS No.: 10241-05-1
Molecular Formula: MoCl5
Formula Weight: 273.24
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: Black
Odour: Pungent
Melting Point: 194°
UN No.: 2508
MDL No.: MFCD00003467
Vzorec:
Cl₅Mo Bod varu: 268 °C (1013 hPa) Bod tání: 194 °C Hustota: 2,928 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003467 CAS číslo: 10241-05-1 EINECS: 233-575-3 UN: 2508 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...