Tisk…

Vanadium(III) acetylacetonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

227110-50G 227110-10G
SIAL227110-50GEA 5330 CZK
SIAL227110-50G SIAL227110-10G
Vanadium(III) acetylacetonate, Sigma-Aldrich®
Vanadium(III) acetylacetonate
Vzorec: C₁₅H₂₁O₆V
Bod varu: 170 °C (0,05 mmHg)
Bod tání: 185…190 °C
Hustota: ∼1 g/cm³ (20 °C)
MDL Number: MFCD00000033
CAS číslo: 13476-99-8
EINECS: 236-759-1
UN: 3285
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR