Tisk…

Vanadium carbide ≥99% (metals basis)

Dodavatel: Thermo Scientific
Varování

12141.14 12141.22
12141.14EA 2280 CZK
12141.14 12141.22
Vanadium carbide ≥99% (metals basis)
Vanadium carbide

Purity: 99% CAS No.: 12070-10-9 Molecular Formula: VC (VC0.88 by X-Ray) Formula Weight: 62.95 Physical Form: -325 Mesh Powder Odour: Odourless UN No.: 3178 MDL No.: MFCD00049698

Vzorec: CV
MW: 62,95 g/mol
Bod varu: 3900 °C (1013 hPa)
Bod tání: 2810 °C
Hustota: 5,77 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049698
CAS číslo: 12070-10-9
EINECS: 235-122-5
UN: 3178
ADR: 4.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR