Tantalum carbide ≥99.5% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Tantalum(IV) carbide
12144.09
12144.18
12144.30
12144.09EA
2550
CZK
12144.09
12144.18
12144.30
Tantalum carbide ≥99.5% (metals basis)
Tantalum carbide
Purity: 99.5%
CAS No.: 12070-06-3
Molecular Formula: TaC
Formula Weight: 192.96
Physical Form: -325 Mesh Powder
Density: 13.9
Appearance: Grey to black
Odour: Odourless
Melting Point: 3880°
Boiling Point: 5500°
UN No.: 3178
MDL No.: MFCD00011255
Vzorec:
CTa Bod varu: 5500 °C Bod tání: 3880 °C Hustota: 13,9 Storage Temperature: Ambient |
MDL Number:
MFCD00011255 CAS číslo: 12070-06-3 EINECS: 235-118-3 UN: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...