Titanium carbide, Sigma-Aldrich®
Dodavatel: Merck
636967-250G
636967-25G
SIAL636967-250GEA
50680
CZK
SIAL636967-250G
SIAL636967-25G
Titanium carbide, Sigma-Aldrich®
Titanium carbide
Vzorec:
CTi MW: 59.89 Bod varu: 4300 Bod tání: 3067 Hustota: 4.93 g/cm³ (25 °C) |
MDL Number:
MFCD00011268 CAS číslo: 12070-08-5 EINECS: 235-120-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...