Tisk…

Titanium carbide ≥99.5% (metals basis), powder

Dodavatel: Thermo Fisher Scientific
Varování

40178.18 40178.30 40178.A1
40178.18EA 2380 CZK
40178.18 40178.30 40178.A1
Titanium carbide ≥99.5% (metals basis), powder
Titanium carbide

Purity: 99.5% CAS No.: 12070-08-5 Molecular Formula: CTi Formula Weight: 59.91 Physical Form: Typically 2 Micron Powder Density: 4.93 Appearance: Dark grey Melting Point: ca 3140° Boiling Point: 4820° UN No.: 3178 MDL No.: MFCD00011268

Vzorec: CTi
MW: 59.89
Bod varu: 4820 °C
Bod tání: ∼ 3140 °C
Hustota: 4.93
Storage Temperature: Ambient
MDL Number: MFCD00011268
CAS číslo: 12070-08-5
EINECS: 235-120-4
UN: 3178
ADR: 4.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR