Silver(I) hexafluorophosphate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Silver hexafluorophosphate
M-Clarity™ quality level = MQ100
227722-10G
227722-1G
SIAL227722-10GEA
8320
CZK
SIAL227722-10G
SIAL227722-1G
Silver(I) hexafluorophosphate, Sigma-Aldrich®
Silver(I) hexafluorophosphate
Vzorec:
AgF₆P MW: 252,83 g/mol |
MDL Number:
MFCD00003415 CAS číslo: 26042-63-7 EINECS: 247-428-6 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...