Tisk…

Copper(II) acetylacetonate, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate

M-Clarity™ quality level = MQ200

C87851-100G C87851-25G C87851-500G
SIALC87851-100GEA 2130 CZK
SIALC87851-100G SIALC87851-25G SIALC87851-500G
Copper(II) acetylacetonate, Sigma-Aldrich®
Copper(II) acetylacetonate
Vzorec: C₁₀H₁₄CuO₄
MW: 261,76 g/mol
Bod tání: 279…283 °C
MDL Number: MFCD00000016
CAS číslo: 13395-16-9
EINECS: 236-477-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR