Copper(II) acetylacetonate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate
M-Clarity™ quality level = MQ100
514365-10G
514365-50G
SIAL514365-10GEA
2140
CZK
SIAL514365-10G
SIAL514365-50G
Copper(II) acetylacetonate, Sigma-Aldrich®
Copper(II) acetylacetonate
Vzorec:
C₁₀H₁₄CuO₄ Bod tání: 279…283 °C |
MDL Number:
MFCD00000016 CAS číslo: 13395-16-9 EINECS: 236-477-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...